Tisk…

3-Oxopentanoic acid 98%

Dodavatel: Apollo Scientific

OR956096-1G OR956096-5G
APOSOR956096-1GEA 3770 CZK
APOSOR956096-1G APOSOR956096-5G
3-Oxopentanoic acid 98%
3-Oxopentanoic acid
Vzorec: C₅H₈O₃
Storage Temperature: Ambient
MDL Number: MFCD11040434
CAS číslo: 10191-25-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR