Tisk…

3-(1,2,4-Oxadiazol-3-yl)pyridine 98%

Dodavatel: Apollo Scientific

OR954508-1G
APOSOR954508-1GEA 9100 CZK
APOSOR954508-1G
3-(1,2,4-Oxadiazol-3-yl)pyridine 98%
3-(1,2,4-Oxadiazol-3-yl)pyridine
Vzorec: C₇H₅N₃O
Storage Temperature: Ambient
MDL Number: MFCD00465052
CAS číslo: 10550-16-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR