Tisk…

2-(5-Oxazolyl)benzonitrile 95%

Dodavatel: Apollo Scientific

OR953406-1G
APOSOR953406-1GEA 8700 CZK
APOSOR953406-1G
2-(5-Oxazolyl)benzonitrile 95%
2-(5-Oxazolyl)benzonitrile
Vzorec: C₁₀H₆N₂O
Storage Temperature: Ambient
MDL Number: MFCD18375279
CAS číslo: 1186127-13-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR