Tisk…

(3-Chlorophenyl)acetic acid ethyl ester 98%

Dodavatel: Apollo Scientific

Synonyms: Ethyl (3-chlorophenyl)acetate, Ethyl 2-(3-chlorophenyl)acetate

OR953372-25G
APOSOR953372-25GEA 2730 CZK
APOSOR953372-25G
(3-Chlorophenyl)acetic acid ethyl ester 98%
(3-Chlorophenyl)acetic acid ethyl ester
Vzorec: (C₆H₄Cl)CH₂COOCH₂CH₃
MW: 198.65
Storage Temperature: Ambient
MDL Number: MFCD07787498
CAS číslo: 14062-29-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR