Tisk…

1-(3-Methoxyphenyl)ethanol 97%

Dodavatel: Apollo Scientific

OR953341-25G OR953341-5G
APOSOR953341-25GEA 8870 CZK
APOSOR953341-25G APOSOR953341-5G
1-(3-Methoxyphenyl)ethanol 97%
1-(3-Methoxyphenyl)ethanol
Vzorec: C₉H₁₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00021866
CAS číslo: 23308-82-9

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR