Tisk…

2-Methyl-1-(2-pyridyl)-1-propylamine 95%

Dodavatel: Apollo Scientific

OR953177-1G OR953177-250MG OR953177-5G
APOSOR953177-1GEA 1910 CZK
APOSOR953177-1G APOSOR953177-250MG APOSOR953177-5G
2-Methyl-1-(2-pyridyl)-1-propylamine 95%
2-Methyl-1-(2-pyridyl)-1-propylamine
Vzorec: C₉H₁₄N₂
Storage Temperature: Refrigerator
MDL Number: MFCD09802325
CAS číslo: 58088-72-5
UN: 2810
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR