Tisk…

Methyl 4-methyl-2-oxopentanoate 95%

Dodavatel: Apollo Scientific

OR953120-1G OR953120-250MG
APOSOR953120-1GEA 12400 CZK
APOSOR953120-1G APOSOR953120-250MG
Methyl 4-methyl-2-oxopentanoate 95%
Methyl 4-methyl-2-oxopentanoate
Vzorec: C₇H₁₂O₃
Storage Temperature: Ambient
MDL Number: MFCD11035886
CAS číslo: 3682-43-7

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR