Tisk…

(3-Cyanophenoxy)acetic acid 98%

Dodavatel: Apollo Scientific

Synonyms: (3-Cyanophenoxy)acetic Acid

OR952746-1G OR952746-5G
APOSOR952746-1GEA 1450 CZK
APOSOR952746-1G APOSOR952746-5G
(3-Cyanophenoxy)acetic acid 98%
(3-Cyanophenoxy)acetic acid
Vzorec: C₉H₇NO₃
MW: 177.157 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00516888
CAS číslo: 1879-58-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR