Tisk…

1-(3-Chlorophenyl)ethanamine 98%

Dodavatel: Apollo Scientific

Synonyms: 1-(3-Chlorophenyl)ethan-1-amine

OR951752-5G
APOSOR951752-5GEA 7870 CZK
APOSOR951752-5G
1-(3-Chlorophenyl)ethanamine 98%
1-(3-Chlorophenyl)ethanamine
Vzorec: C₈H₁₀ClN
MW: 155.63 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05215231
CAS číslo: 24358-43-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR