Tisk…

2-(2-Cyanophenoxy)acetic acid 98%

Dodavatel: Apollo Scientific

Synonyms: (2-Cyanophenoxy)acetic Acid

OR951075-1G OR951075-25G OR951075-5G
APOSOR951075-1GEA 622 CZK
APOSOR951075-1G APOSOR951075-25G APOSOR951075-5G
2-(2-Cyanophenoxy)acetic acid 98%
2-(2-Cyanophenoxy)acetic acid
Vzorec: C₉H₇NO₃
MW: 177.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02257373
CAS číslo: 6574-95-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR