Tisk…

trans-Methyl 3-hydroxy-1-methyl-cyclobutanecarboxylate

Dodavatel: Apollo Scientific

OR949332-250MG
APOSOR949332-250MGEA 16500 CZK
APOSOR949332-250MG
trans-Methyl 3-hydroxy-1-methyl-cyclobutanecarboxylate
trans-Methyl 3-hydroxy-1-methyl-cyclobutanecarboxylate
Vzorec: C₇H₁₂O₃
Storage Temperature: Ambient
MDL Number: MFCD23105988
CAS číslo: 1408075-48-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR