Tisk…

5-(2-Methylphenyl)-1,3,4-oxadiazol-2-amine 95%

Dodavatel: Apollo Scientific

OR946701-1G
APOSOR946701-1GEA 8270 CZK
APOSOR946701-1G
5-(2-Methylphenyl)-1,3,4-oxadiazol-2-amine 95%
5-(2-Methylphenyl)-1,3,4-oxadiazol-2-amine
Vzorec: C₉H₉N₃O
Storage Temperature: Ambient
MDL Number: MFCD00652824
CAS číslo: 1750-78-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR