Tisk…

1-Boc-2-acetyl-pyrrolidine 97%

Dodavatel: Apollo Scientific

OR944671-1G OR944671-500MG
APOSOR944671-1GEA 13390 CZK
APOSOR944671-1G APOSOR944671-500MG
1-Boc-2-acetyl-pyrrolidine 97%
1-Boc-2-acetyl-pyrrolidine
Vzorec: C₁₁H₁₉NO₃
Storage Temperature: Ambient
MDL Number: MFCD16308624
CAS číslo: 92012-22-1

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR