Tisk…

(tert-Butyldimethylsiloxy)acetaldehyde 95%

Dodavatel: Apollo Scientific

OR941768-5G
APOSOR941768-5GEA 1820 CZK
APOSOR941768-5G
(tert-Butyldimethylsiloxy)acetaldehyde 95%
(tert-Butyldimethylsiloxy)acetaldehyde
Vzorec: C₈H₁₈O₂Si
Bod varu: 165…167 °C
Hustota: 0,915 g/cm³ (25 °C)
Bod vzplanutí: 60 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01321229
CAS číslo: 102191-92-4
UN: 1989
ADR: 3,III

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR