Tisk…

1-Methyl-1H-indol-2-amine hydrochloride 95%

Dodavatel: Apollo Scientific

OR941089-1G OR941089-250MG
APOSOR941089-1GEA 17990 CZK
APOSOR941089-1G APOSOR941089-250MG
1-Methyl-1H-indol-2-amine hydrochloride 95%
1-Methyl-1H-indol-2-amine hydrochloride
Vzorec: C₉H₁₀N₂·ClH
Storage Temperature: Ambient
MDL Number: MFCD15143632
CAS číslo: 42456-82-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR