Tisk…

Ethyl tetrahydrofuran-2-acetate 95%

Dodavatel: Apollo Scientific

OR939909-10G OR939909-1G OR939909-5G
APOSOR939909-10GEA 22950 CZK
APOSOR939909-10G APOSOR939909-1G APOSOR939909-5G
Ethyl tetrahydrofuran-2-acetate 95%
Ethyl tetrahydrofuran-2-acetate
Vzorec: C₈H₁₄O₃
Storage Temperature: Ambient
MDL Number: MFCD00059735
CAS číslo: 2434-02-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR