Tisk…

4-Methoxyphenethyl bromide 97%

Dodavatel: Apollo Scientific

Synonyms: 1-Bromo-2-(4-methoxyphenyl)ethane, 1-(2-Bromoethyl)-4-methoxybenzene, 4-(2-Bromoethyl)anisole

OR939726-5G
APOSOR939726-5GEA 1910 CZK
APOSOR939726-5G
4-Methoxyphenethyl bromide 97%
4-Methoxyphenethyl bromide
Vzorec: C₉H₁₁BrO
MW: 215.09
Storage Temperature: Ambient
MDL Number: MFCD07369786
CAS číslo: 14425-64-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR