Tisk…

1-Cyclopropyl-2-nitroethanol 95%

Dodavatel: Apollo Scientific

OR939529-1G
APOSOR939529-1GEA 17990 CZK
APOSOR939529-1G
1-Cyclopropyl-2-nitroethanol 95%
1-Cyclopropyl-2-nitroethanol
Vzorec: C₅H₉NO₃
Storage Temperature: Ambient
MDL Number: MFCD20484062
CAS číslo: 54120-03-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR