Tisk…

N-tert-Butyltrimethylsilylamine 97%

Dodavatel: Apollo Scientific

OR939298-100G OR939298-25G
APOSOR939298-100GEA 13630 CZK
APOSOR939298-100G APOSOR939298-25G
N-tert-Butyltrimethylsilylamine 97%
N-tert-Butyltrimethylsilylamine
Vzorec: C₇H₁₉NSi
Storage Temperature: Ambient
MDL Number: MFCD00054891
CAS číslo: 5577-67-3

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR