Tisk…

trans-4-(Hydroxymethyl)cyclohexanol 95%

Dodavatel: Apollo Scientific

OR938871-1G OR938871-250MG
APOSOR938871-1GEA 2940 CZK
APOSOR938871-1G APOSOR938871-250MG
trans-4-(Hydroxymethyl)cyclohexanol 95%
trans-4-(Hydroxymethyl)cyclohexanol
Vzorec: C₇H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD11036248
CAS číslo: 3685-27-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR