Tisk…

1-Phenylethyl acetate 95%

Dodavatel: Apollo Scientific

Synonyms: Styrallyl acetate, α-Methylbenzyl acetate

OR938810-500G
APOSOR938810-500GEA 2240 CZK
APOSOR938810-500G
1-Phenylethyl acetate 95%
1-Phenylethyl acetate
Vzorec: C₁₀H₁₂O₂
MW: 164.2
Bod varu: 94
Storage Temperature: Ambient
MDL Number: MFCD00008858
CAS číslo: 93-92-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR