Tisk…

3-Bromo-3-methyl-2-butanone 96%

Dodavatel: Apollo Scientific

OR938435-25G OR938435-5G
APOSOR938435-25GEA 13030 CZK
APOSOR938435-25G APOSOR938435-5G
3-Bromo-3-methyl-2-butanone 96%
3-Bromo-3-methyl-2-butanone
Vzorec: C₅H₉BrO
MDL Number: MFCD06661470
CAS číslo: 2648-71-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR