Tisk…

Ethyl-2-methyl-3-furancarboxylate 95%

Dodavatel: Apollo Scientific

OR938025-1G OR938025-5G
APOSOR938025-1GEA 4140 CZK
APOSOR938025-1G APOSOR938025-5G
Ethyl-2-methyl-3-furancarboxylate 95%
Ethyl-2-methyl-3-furancarboxylate
Vzorec: C₈H₁₀O₃
Storage Temperature: Ambient
MDL Number: MFCD00134327
CAS číslo: 28921-35-9

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR