Tisk…

3,4-Dimethylcyclohexanone 95%

Dodavatel: Apollo Scientific

OR938021-10G OR938021-1G OR938021-5G
APOSOR938021-10GEA 28300 CZK
APOSOR938021-10G APOSOR938021-1G APOSOR938021-5G
3,4-Dimethylcyclohexanone 95%
3,4-Dimethylcyclohexanone
Vzorec: C₈H₁₄O
Storage Temperature: Ambient
MDL Number: MFCD00045552
CAS číslo: 5465-09-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR