Tisk…

Tetrahydrofurfuryl propionate 97%

Dodavatel: Apollo Scientific

Synonyms: (Tetrahydrofuran-2-yl)methyl propionate

OR937571-100G OR937571-25G OR937571-500G
APOSOR937571-100GEA 2780 CZK
APOSOR937571-100G APOSOR937571-25G APOSOR937571-500G
Tetrahydrofurfuryl propionate 97%
Tetrahydrofurfuryl propionate
Vzorec: C₈H₁₄O₃
MW: 158.1950 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00047080
CAS číslo: 637-65-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR