Tisk…

1-(3-Bromopropyl)pyrrole 95%

Dodavatel: Apollo Scientific

OR937517-1G OR937517-250MG
APOSOR937517-1GEA 7080 CZK
APOSOR937517-1G APOSOR937517-250MG
1-(3-Bromopropyl)pyrrole 95%
1-(3-Bromopropyl)pyrrole
Vzorec: C₇H₁₀BrN
Storage Temperature: Ambient
MDL Number: MFCD00191324
CAS číslo: 100779-91-7

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR