Tisk…

4-Isopropylcyclohexylamine (cis- and trans- mixture) 95%

Dodavatel: Apollo Scientific

OR937443-1G OR937443-5G
APOSOR937443-1GEA 4770 CZK
APOSOR937443-1G APOSOR937443-5G
4-Isopropylcyclohexylamine (cis- and trans- mixture) 95%
4-Isopropylcyclohexylamine (cis- and trans- mixture)
Vzorec: C₉H₁₉N
Storage Temperature: Ambient
MDL Number: MFCD16853065
CAS číslo: 52430-81-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR