Tisk…

4,4-Dimethyl-2-imidazolidinethione 95%

Dodavatel: Apollo Scientific

Synonyms: 2-Mercapto-4,4-dimethylimidazoline, 4,4-Dimethylethylenethiourea

OR937418-5G
APOSOR937418-5GEA 5290 CZK
APOSOR937418-5G
4,4-Dimethyl-2-imidazolidinethione 95%
4,4-Dimethyl-2-imidazolidinethione
Vzorec: C₅H₁₀N₂S
MW: 130.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00661072
CAS číslo: 6086-42-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR