Tisk…

Ethylene glycol dimethacrylate (ethylene dimethacrylate) 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: Ethylene dimethacrylate, Ethane-1,2-diyl bis(2-methylacrylate)

OR936412-25G OR936412-100G
APOSOR936412-25GEA 996 CZK
APOSOR936412-25G APOSOR936412-100G
Ethylene glycol dimethacrylate (ethylene dimethacrylate) 95%
Ethylene glycol dimethacrylate (ethylene dimethacrylate)
Vzorec: C₁₀H₁₄O₄
MW: 198.21 g/mol
Bod varu: 98…100 °C
Bod tání: –40 °C
Bod vzplanutí: 104 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008590
CAS číslo: 97-90-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR