Tisk…

Phenylsuccinic anhydride 95%

Dodavatel: Apollo Scientific

Synonyms: 3-Phenyldihydrofuran-2,5-dione

OR936348-25G
APOSOR936348-25GEA 3060 CZK
APOSOR936348-25G
Phenylsuccinic anhydride 95%
Phenylsuccinic anhydride
Vzorec: C₁₀H₈O₃
MW: 176.1700 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00192395
CAS číslo: 1131-15-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR