Tisk…

(S)-(-)-1-Methyl-2-(1-piperidinomethyl)pyrrolidine 95%

Dodavatel: Apollo Scientific

Synonyms: (S)-(-)-1-[(1-Methyl-2-pyrrolidinyl)methyl]piperidine

OR936287-1G
APOSOR936287-1GEA 2440 CZK
APOSOR936287-1G
(S)-(-)-1-Methyl-2-(1-piperidinomethyl)pyrrolidine 95%
(S)-(-)-1-Methyl-2-(1-piperidinomethyl)pyrrolidine
Vzorec: C₁₁H₂₂N₂
MW: 182.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00077821
CAS číslo: 84466-85-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR