Tisk…

3-Chloro-N-methylpyridin-2-amine 95%

Dodavatel: Apollo Scientific

OR932541-1G OR932541-5G
APOSOR932541-1GEA 9330 CZK
APOSOR932541-1G APOSOR932541-5G
3-Chloro-N-methylpyridin-2-amine 95%
3-Chloro-N-methylpyridin-2-amine
Vzorec: C₆H₇ClN₂
Storage Temperature: Ambient
MDL Number: MFCD11122502
CAS číslo: 468718-67-4

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR