Tisk…

Methyl-4-Isopropoxybenzoate 97%

Dodavatel: Apollo Scientific

Synonyms: 4-Isopropoxybenzoic Acid Methyl Ester, Methyl 4-isopropoxybenzoate

OR931955-5G
APOSOR931955-5GEA 2900 CZK
APOSOR931955-5G
Methyl-4-Isopropoxybenzoate 97%
Methyl-4-Isopropoxybenzoate
Vzorec: C₁₁H₁₄O₃
MW: 194,23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00084855
CAS číslo: 35826-59-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR