Tisk…

3-Butoxypropionitrile 95%

Dodavatel: Apollo Scientific

OR931713-100G
APOSOR931713-100GEA 2030 CZK
APOSOR931713-100G
3-Butoxypropionitrile 95%
3-Butoxypropionitrile
Vzorec: C₇H₁₃NO
Storage Temperature: Ambient
MDL Number: MFCD00045643
CAS číslo: 6959-71-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR