Tisk…

1-(4-Methyl-2-(methylthio)pyrimidin-5-yl)ethanone 95%

Dodavatel: Apollo Scientific

OR931619-100MG OR931619-1G OR931619-250MG
APOSOR931619-100MGEA 3310 CZK
APOSOR931619-100MG APOSOR931619-1G APOSOR931619-250MG
1-(4-Methyl-2-(methylthio)pyrimidin-5-yl)ethanone 95%
1-(4-Methyl-2-(methylthio)pyrimidin-5-yl)ethanone
Vzorec: C₈H₁₀N₂OS
Storage Temperature: Ambient
MDL Number: MFCD00223551
CAS číslo: 66373-26-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR