Tisk…

3-[1-(Dimethylamino)ethyl]benzaldehyde 95%

Dodavatel: Apollo Scientific

OR931608-1G OR931608-250MG
APOSOR931608-1GEA 11380 CZK
APOSOR931608-1G APOSOR931608-250MG
3-[1-(Dimethylamino)ethyl]benzaldehyde 95%
3-[1-(Dimethylamino)ethyl]benzaldehyde
Vzorec: C₁₁H₁₅NO
Storage Temperature: Ambient
MDL Number: MFCD08059975
CAS číslo: 915924-91-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR