Tisk…

6-Methyl-1,2,3,4-tetrahydro-isoquinoline hydrochloride 95%

Dodavatel: Apollo Scientific

OR931101-1G OR931101-250MG
APOSOR931101-1GEA 9530 CZK
APOSOR931101-1G APOSOR931101-250MG
6-Methyl-1,2,3,4-tetrahydro-isoquinoline hydrochloride 95%
6-Methyl-1,2,3,4-tetrahydro-isoquinoline hydrochloride
Vzorec: C₁₀H₁₃N·ClH
Storage Temperature: Ambient
MDL Number: MFCD09832197
CAS číslo: 42923-76-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR