Tisk…

FOR-NLE-OH 95%

Dodavatel: Apollo Scientific

OR930495-1G OR930495-250MG
APOSOR930495-1GEA 6220 CZK
APOSOR930495-1G APOSOR930495-250MG
FOR-NLE-OH 95%
FOR-NLE-OH
Vzorec: C₇H₁₃NO₃
MW: 159.185 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00237678
CAS číslo: 133388-96-2

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR