Tisk…

tert-Butyl N-(1H-pyrazol-4-ylmethyl)carbamate 95%

Dodavatel: Apollo Scientific

OR930419-100MG OR930419-1G OR930419-250MG
APOSOR930419-100MGEA 3710 CZK
APOSOR930419-100MG APOSOR930419-1G APOSOR930419-250MG
tert-Butyl N-(1H-pyrazol-4-ylmethyl)carbamate 95%
tert-Butyl N-(1H-pyrazol-4-ylmethyl)carbamate
Vzorec: C₉H₁₅N₃O₂
Storage Temperature: Ambient
MDL Number: MFCD12820298
CAS číslo: 1107620-72-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR