Tisk…

6-tert-Butylnicotinic acid 98%

Dodavatel: Apollo Scientific

Synonyms: 6-(tert-Butyl)nicotinic acid

OR930175-1G
APOSOR930175-1GEA 1780 CZK
APOSOR930175-1G
6-tert-Butylnicotinic acid 98%
6-tert-Butylnicotinic acid
Vzorec: C₁₀H₁₃NO₂
MW: 179.216 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08059362
CAS číslo: 832715-99-8

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR