Tisk…

D-Valine ethyl ester hydrochloride 95%

Dodavatel: Apollo Scientific

Synonyms: H-D-Val-OEt·HCl, (R)-Ethyl 2-amino-3-methylbutanoate hydrochloride

OR929780-5G
APOSOR929780-5GEA 1490 CZK
APOSOR929780-5G
D-Valine ethyl ester hydrochloride 95%
D-Valine ethyl ester hydrochloride
Vzorec: C₇H₁₅NO₂·HCl
MW: 181.66 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00191043
CAS číslo: 73913-64-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR