Tisk…

Sulphachloropyrazine sodium 98%

Dodavatel: Apollo Scientific

OR928389-1G OR928389-5G
APOSOR928389-1GEA 8070 CZK
APOSOR928389-1G APOSOR928389-5G
Sulphachloropyrazine sodium 98%
Sulphachloropyrazine sodium
Vzorec: C₁₀H₈ClN₄NaO₂S
Storage Temperature: Ambient
MDL Number: MFCD06798154
CAS číslo: 71720-40-6

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR