Tisk…

4,4-Dimethoxybutanenitrile 95%

Dodavatel: Apollo Scientific

OR928315-25G OR928315-100G
APOSOR928315-25GEA 3580 CZK
APOSOR928315-25G APOSOR928315-100G
4,4-Dimethoxybutanenitrile 95%
4,4-Dimethoxybutanenitrile
Vzorec: C₆H₁₁NO₂
Storage Temperature: Ambient
MDL Number: MFCD00019883
CAS číslo: 14618-78-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR