Tisk…

4-Acetyl-L-phenylalanine 97%

Dodavatel: Apollo Scientific

Synonyms: H-Phe(4-Ac)-OH

OR928215-1G
APOSOR928215-1GEA 11480 CZK
APOSOR928215-1G
4-Acetyl-L-phenylalanine 97%
4-Acetyl-L-phenylalanine
Vzorec: C₁₁H₁₃NO₃
MW: 207.226 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06659983
CAS číslo: 122555-04-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR