Tisk…

Methyl-o-anisate 95%

Dodavatel: Apollo Scientific

OR927757-500G
APOSOR927757-500GEA 5720 CZK
APOSOR927757-500G
Methyl-o-anisate 95%
Methyl-o-anisate
Vzorec: C₉H₁₀O₃
Storage Temperature: Ambient
MDL Number: MFCD00008423
CAS číslo: 606-45-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR