Tisk…

(2,5-Dimethoxyphenyl)acetic acid 95%

Dodavatel: Apollo Scientific

OR927331-100G OR927331-25G OR927331-5G
APOSOR927331-100GEA 22290 CZK
APOSOR927331-100G APOSOR927331-25G APOSOR927331-5G
(2,5-Dimethoxyphenyl)acetic acid 95%
(2,5-Dimethoxyphenyl)acetic acid
Vzorec: (C₆H₃(OCH₃)₂)CH₂COOH
Storage Temperature: Ambient
MDL Number: MFCD00004322
CAS číslo: 1758-25-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR