Tisk…

2-(4-Methylpiperazin-1-ylmethyl)benzylamine 97%

Dodavatel: Apollo Scientific

OR9268-1G OR9268-5G
APOSOR9268-1GEA 7050 CZK
APOSOR9268-1G APOSOR9268-5G
2-(4-Methylpiperazin-1-ylmethyl)benzylamine 97%
2-(4-Methylpiperazin-1-ylmethyl)benzylamine
Vzorec: C₁₃H₂₁N₃
Storage Temperature: Ambient
CAS číslo: 879896-50-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR