Tisk…

3-(Chloromethyl)-5-isopropyl-1,2,4-oxadiazole 95%

Dodavatel: Apollo Scientific

OR925985-1G OR925985-250MG
APOSOR925985-1GEA 22950 CZK
APOSOR925985-1G APOSOR925985-250MG
3-(Chloromethyl)-5-isopropyl-1,2,4-oxadiazole 95%
3-(Chloromethyl)-5-isopropyl-1,2,4-oxadiazole
Vzorec: C₆H₉ClN₂O
Storage Temperature: Ambient
MDL Number: MFCD09864313
CAS číslo: 189130-87-8

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR