Tisk…

Methyl cis-4-aminocyclohexanecarboxylate hydrochloride 96%

Dodavatel: Apollo Scientific

OR925859-5G
APOSOR925859-5GEA 3740 CZK
APOSOR925859-5G
Methyl cis-4-aminocyclohexanecarboxylate hydrochloride 96%
Methyl cis-4-aminocyclohexanecarboxylate hydrochloride
Vzorec: C₈H₁₅NO₂
Storage Temperature: Ambient
MDL Number: MFCD08274537
CAS číslo: 61367-16-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR