Tisk…

4-tert-Butylpyridine 95%

Dodavatel: Apollo Scientific

Synonyms: 4-(tert-Butyl)pyridine

OR924555-25G
APOSOR924555-25GEA 1700 CZK
APOSOR924555-25G
4-tert-Butylpyridine 95%
4-tert-Butylpyridine
Vzorec: C₉H₁₃N
MW: 135.21
Bod varu: 196
Bod tání: -0.9
Hustota: 0.9219
Storage Temperature: Ambient
MDL Number: MFCD00006435
CAS číslo: 3978-81-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR